Would you like to log in again?
Washed Pooled Cells, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R403-0050 R400-0100 R407-0100 R407-0100 R400-0050 R402-0100 R402-0050 R402-0100 R402-0050 R406-0100 R403-0100 R407-0050 R401-0050 R401-0050 R407-0050 R406-0100 R405-0050 R404-0100 R401-0100 R404-0100 R405-0050 R401-0100 R405-0100 R406-0050 R404-0050 R400-0100 R405-0100 R403-0050 R406-0050 R400-0050 R403-0100 R404-0050
RLR404-0050EA 458 USD InStock
CAR400-0050 RLR404-0050 RLR407-0050 CAR406-0050 CAR403-0050 RLR405-0100 RLR400-0100 CAR404-0100 CAR401-0100 RLR402-0100 RLR406-0100 RLR403-0100 CAR402-0100 RLR401-0050 CAR405-0100 CAR407-0050 RLR404-0100 RLR401-0100 CAR406-0100 CAR400-0100 RLR405-0050 CAR404-0050 CAR401-0050 RLR407-0100 CAR403-0100 RLR402-0050 CAR402-0050 RLR403-0050 CAR405-0050 RLR400-0050 CAR407-0100 RLR406-0050
Washed Pooled Cells, Rockland Immunochemicals
Blood
Suspensions are suitable for complement fixation, hemagglutination, hemadsorption, and other related serological procedures
All suspensions are 10% cell solutions in saline, except for RLR406-0050 and RLR406-0100. These two suspensions are 100% cells. Suspensions contain no stabilizers or preservatives.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty