Would you like to log in again?
TURKEY RED BLOOD CELLS 10% Washed Pooled Cells, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R408-0050 R408-0100
RLR408-0050EA 507.54 USD InStock
RLR408-0050 RLR408-0100
TURKEY RED BLOOD CELLS 10% Washed Pooled Cells, Rockland Immunochemicals
Blood
TURKEY RED BLOOD CELLS 10% Washed Pooled Cells
  • aseptically collected and prepared to order
  • Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS)
Turkey whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Turkey red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Turkey red blood cells are perishable and are collected and processed upon receipt of your order.
Complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty