Would you like to log in again?
Rat Tissue, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
RT-T075 RT-T077 RT-T103 RT-T090 RT-T087 RT-T089 RT-T098 RT-T074 RT-T097 RT-T107 RT-T088 RT-T093 RT-T074 RT-T093 RT-T086 RT-T087 RT-T077 RT-T101 RT-T098 RT-T110 RT-T081 RT-T085 RT-T110 RT-T083 RT-T105 RT-T108 RT-T109 RT-T076 RT-T095 RT-T108 RT-T083 RT-T105 RT-T085 RT-T089 RT-T075 RT-T109 RT-T084 RT-T090 RT-T096 RT-T097 RT-T095 RT-T106 RT-T104 RT-T086 RT-T096 RT-T094 RT-T084 RT-T101 RT-T102 RT-T081 RT-T102 RT-T106 RT-T094 RT-T103 RT-T076 RT-T107 RT-T104 RT-T088
RLRT-T075EA 62 USD InStock
CART-T076 CART-T077 CART-T110 CART-T074 CART-T075 RLRT-T075 RLRT-T074 RLRT-T077 RLRT-T110 RLRT-T076 CART-T087 CART-T088 CART-T089 CART-T083 RLRT-T105 CART-T084 RLRT-T104 RLRT-T103 CART-T085 CART-T086 RLRT-T102 RLRT-T109 RLRT-T108 RLRT-T107 CART-T081 RLRT-T106 RLRT-T101 CART-T098 CART-T094 CART-T095 CART-T096 CART-T097 CART-T090 CART-T093 RLRT-T097 RLRT-T096 RLRT-T095 RLRT-T094 RLRT-T098 RLRT-T093 RLRT-T090 CART-T102 CART-T103 CART-T104 CART-T105 CART-T101 RLRT-T086 RLRT-T085 RLRT-T084 RLRT-T083 RLRT-T089 RLRT-T088 RLRT-T087 CART-T106 RLRT-T081 CART-T107 CART-T108 CART-T109
Rat Tissue, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Use for DNA and RNA isolation.
  • Collected from antibiotic-free animals
  • DNA/RNA isolation
All tissues are collected from normal, healthy, mature animals that have been maintained on an antibiotic-free diet. Tissues are stored at −20°C or colder prior to use.
Ordering information: For specific breed, sex, or weight requirements, please contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty