Would you like to log in again?
Cell Lysates, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
W09-000-365 W09-000-361 W09-000-364 W09-000-363 W09-000-364 W10-001-369 W09-000-366 W09-000-362 W09-001-374 W10-001-369 W09-001-370 W10-000-358 W09-000-360 W09-001-370 W09-001-369 W09-000-360 W09-000-361 W09-001-369 W09-001-374 W09-000-362 W09-000-366 W10-000-358 W09-000-363 W09-001-368 W09-000-365 W09-001-367 W10-001-371 W09-001-367 W10-001-371 W09-001-368
RLW10-001-369EA 161.51 USD InStock
CAW09-000-361 CAW09-000-362 CAW09-000-360 RLW10-001-369 CAW09-000-365 CAW09-000-366 CAW09-000-363 CAW09-000-364 CAW10-001-371 RLW09-001-367 RLW09-001-368 RLW09-001-369 CAW10-001-369 RLW10-000-358 CAW09-001-367 CAW09-001-368 CAW09-001-369 CAW10-000-358 RLW10-001-371 RLW09-001-370 RLW09-001-374 CAW09-001-370 RLW09-000-361 CAW09-001-374 RLW09-000-362 RLW09-000-360 RLW09-000-365 RLW09-000-366 RLW09-000-363 RLW09-000-364
Cell Lysates, Rockland Immunochemicals
Cells
Ideal as positive controls for Western blotting, ready-to-use cell lysates are derived from cell lines using highly refined extraction protocols to ensure quality, protein integrity, and lot-to-lot reproducibility
Each vial contains 1mg/mL of protein in SDS-PAGE sample buffer. The sample buffer consists of 0.05mM TRIS HCl, 2% SDS, 10% glycerol, and 0.1% bromophenol blue at pH 6.8. All lysates are tested by SDS-PAGE using 4–20% gradient gels and immunoblot analysis using antibodies to key cell signaling components for confirming the presence of both high and low molecular weight proteins.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty