Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
J.T.Baker®, Syringe Filters
Supplier: Avantor
J.T.Baker®
SF02-94 SF01-36 SF02-78 SF01-92 SF01-60 SF01-18 SF02-36 SF01-70 SF02-50 SF01-16 SF01-06 SF01-02 SF02-74 SF01-22 SF02-08 SF01-96 SF02-68 SF02-58 SF01-34 SF01-52 SF02-88 SF02-04 SF01-28 SF01-88 SF02-28 SF02-76 SF02-48 SF01-90 SF01-44 SF02-82 SF02-22 SF02-10 SF01-48 SF02-06 SF02-34 SF01-94 SF02-12 SF02-14 SF02-44 SF02-46 SF01-42 SF01-64 SF01-40 SF01-30 SF01-58 SF02-02 SF02-86 SF02-80 SF01-04 SF01-78 SF01-38 SF02-54 SF01-74 SF02-40 SF02-60 SF02-26 SF01-68 SF02-20 SF01-24 SF01-84 SF02-32 SF01-32 SF02-90 SF01-26 SF01-08 SF01-62 SF01-10 SF02-70 SF02-42 SF01-80 SF01-46 SF02-18 SF02-62 SF02-52 SF01-72 SF02-92 SF01-98 SF01-50 SF02-64 SF01-76 SF01-14 SF02-84 SF02-24 SF02-38 SF01-12 SF01-54 SF02-16 SF01-82 SF02-30 SF02-72 SF01-20 SF01-86
SF01-82CS 6604.69 USD InStock
SF01-82 SF02-52 SF01-80 SF02-10 SF02-54 SF01-42 SF01-86 SF02-92 SF01-40 SF01-84 SF02-94 SF02-50 SF01-02 SF01-46 SF02-16 SF01-88 SF01-44 SF02-18 SF01-06 SF02-12 SF01-48 SF01-04 SF02-58 SF02-14 SF02-08 SF02-90 SF02-62 SF01-92 SF02-20 SF02-64 SF01-90 SF01-96 SF01-52 SF02-60 SF01-94 SF01-50 SF02-26 SF01-12 SF02-28 SF01-10 SF01-98 SF01-54 SF02-22 SF01-16 SF02-68 SF02-24 SF01-58 SF01-14 SF01-08 SF01-60 SF02-74 SF02-30 SF02-76 SF02-32 SF01-64 SF01-20 SF02-70 SF01-62 SF02-72 SF01-68 SF01-24 SF02-38 SF01-22 SF01-28 SF02-78 SF02-34 SF01-26 SF02-36 SF01-18 SF02-40 SF02-84 SF01-70 SF02-86 SF02-42 SF02-80 SF01-30 SF01-74 SF02-82 SF01-72 SF02-04 SF02-48 SF01-34 SF01-78 SF02-06 SF01-32 SF01-76 SF02-88 SF02-44 SF01-38 SF02-46 SF02-02 SF01-36
J.T.Baker®, Syringe Filters
Filters Syringe Filters
These syringe filters are specifically designed for chromatography sample preparation applications and optimized to provide the most consistent results with less extractables.
  • HPLC certified
  • Biosafe according to Class VI plastics tests
  • Polypropylene housing material
  • Non sterile
  • Each filter has a female Luer-lock inlet and a male slip Luer outlet
  • Large pore size 1,0 µm for high throughput and fast flow rates
Available with PTFE (hydrophilic or hydrophobic), nylon, PES, RC membrane and GF (glass fiber):
- Water wettable hydrophilic PTFE offers excellent chemical resistance to both common aqueous and aggressive organic solvents, most of the acids and alkalis.
- Hydrophobic PTFE resistant to the majority of solvents as well as strong acids and bases, also suitable for gas filtration.
- Nylon provides a wide range of chemical compatibility, suitable for filtration of either aqueous or organic solvents. It is a natural hydrophilic membrane that can be used in a broad pH range.
- PES offers lower protein binding as well as higher flow rate.
- Regenerated cellulose exhibits low protein binding and wide chemical resistance.
- Built in glass fibre (100% binder-free borosilicate glass fiber) prefilter double layer type is specially for high-particle loading, ideal for difficult to filter liquids.
Certifications: Manufactured under ISO9001:2015 quality management.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty