Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EletroTen-Blue Electroporation-Competent Cells, Agilent Technologies
Supplier: Agilent Technologies
200159 200159
ST200159EA 320.71 USD InStock
CAST200159 ST200159
EletroTen-Blue Electroporation-Competent Cells, Agilent Technologies
Competent Cells
Achieve the highest transformation efficiencies available when cloning large plasmids and ligated DNA.
  • Separate DNA fragments and libraries up to 5000 bp in 1 to 2 min/sample
  • Achieve objective quantitative and purity measurements from as little as 2 µl of DNA, sensitive down to 5 pg/µl
  • Ideal for sample QC in NGS workflows
  • Partially used ScreenTape devices can be saved and used at a later time
  • No need to batch samples
Agilent EletroTen-Blue Electroporation-Competent Cells offer the highest transformation efficiencies available, dramatically improving cloning large plasmids and ligated DNA. These cells also offer the highest electroporation efficiency (Hee) phenotype, which confers increased resilience in harsh electroporation conditions.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty