Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
XL10-Gold Ultracompetent Cells
Supplier: Agilent Technologies
200315 200317 200317 200314 200315 200314
ST200317EA 342.16 USD InStock
CAST200314 ST200317 CAST200317 CAST200315 ST200315 ST200314
XL10-Gold Ultracompetent Cells
Competent Cells
XL10-Gold ultracompetent cells transform large plasmids and ligated DNA with highest efficiency.
  • Engineered to transform large plasmids and ligated DNA more efficiently than other cell lines
  • Includes Hte phenotype for 20-fold to 30-fold higher transformation efficiency of ligated DNA and better competent cell performance
  • Chemically competent cells that allow efficient transformation of large DNA molecules, including expression vectors and genomic DNA
  • Ideal for plasmid library construction by decreasing bias against larger plasmids and producing more colonies for a more representative library
  • Transformation efficiency of ≥5×10⁹ transformants/µg
The XL10-Gold ultracompetent cells were designed for cloning large plasmids and ligated DNA with the highest transformation efficiency possible, while exhibiting faster growth and larger colonies. XL10-Gold Ultracompetent Cells possess the high transformation phenotype (Hte), which gives these cells an average transformation efficiency of ≥5×10⁹ transformants/μg of supercoiled DNA Plasmid libraries constructed in this strain are more representative because XL10-Gold cells decrease the bias against large inserts.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty