Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

qPCR Consumables, Agilent Technologies

Supplier: Agilent Technologies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Agilent
410022 401266 410024 410024 401425 410022 401428
ST410024EA 40.26 USD InStock
ST410024 ST401428 CAST410024 CAST410022 ST401425 99900-234 ST410022
qPCR Consumables, Agilent Technologies
Thermal Cyclers
These QPCR Plastics and accessories have been validated for optimal performance with Mx3005P, Mx3000P and the AriaMx real time PCR Systems.

Plates and tubes fit most PCR instruments. Racks hold standard PCR and microcentrifuge tubes.

The StrataCooler LP benchtop cooler provides the most protection for enzymes and the cooler will maintain a temperature measuring as low as –15 °C even after 2 hours.

StrataCooler Cryo preservation modules enable highly controlled freezing (0.4 to 0.6 °C/minute) of mammalian and insect cells to significantly increase survival rates (up to 90%).

96-Well Semi-skirted Polypropylene PCR plate validated for optimal performance and compatible with the optical Strip Caps.

Caution: For research use only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty