Would you like to log in again?
QC Sample pH 7, Calibration Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
133
SX133EA 71.75 USD InStock
SX133
QC Sample pH 7, Calibration Standard, SPEX CertiPrep
Buffers pH Calibration Buffers
ERA Cal pH Buffers are directly traceable to NIST SRMs, mercury-free, guaranteed stable for at least one year after your receipt and are supplied with a full CoA.
The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and methof ruggedness.
QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty