Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Acetate (C₂H₃O₂) Single-Element Ion Anion Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
AS-ACE9-2X AS-ACE9-2Y
SXAS-ACE9-2XEA 171.59 USD InStock
SXAS-ACE9-2X SXAS-ACE9-2Y
Acetate (C₂H₃O₂) Single-Element Ion Anion Standard, SPEX CertiPrep
Standards Anionic Standards Acetate Standards
Ion chromatography and ion selective electrode standards.
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty