Would you like to log in again?
Sulfur Biodiesel Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
BFS-15Y BFS-20Y BFS-50Y BFS-100Y BFM-10Y BFS-10Y BFM-20Y BFS-25Y BFS-5Y BFM-5Y
SXBFS-100YEA 150.86 USD InStock
SXBFS-100Y SXBFS-5Y SXBFS-20Y SXBFS-50Y SXBFM-10Y SXBFM-20Y SXBFS-10Y SXBFS-15Y SXBFM-5Y SXBFS-25Y
Sulfur Biodiesel Standard, SPEX CertiPrep
Standards Petroleum Industry Standards
B100 Biodiesel Standards
Governments worldwide have passed regulations that mandate lower levels of sulfur in biodiesel fuel. To comply with the implementation of these regulations, SPEX CertiPrep offers specifically designed Certified Reference Materials for industrial use.
Certifications: The B100 Biodiesel Standards meet the requirements for testing ASTM Methods D6751, D5453 and EN 14214.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty