Would you like to log in again?
Spex® CertiPrep CAN-TERP-HA High Abundance Terpene Mix
Supplier: SPEX CertiPrep
CAN-TERP-HA
SXCAN-TERP-HAEA 115.84 USD InStock
SXCAN-TERP-HA
Spex® CertiPrep CAN-TERP-HA High Abundance Terpene Mix
Standards Organic Standards
High concentration analytical standard for terpene analysis contains the most abundant terpenes found in cannabis products
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 5,000 ppm high abundance 10 component terpene mix in methanol matrix
  • 1 mL ampule
This Spex CertiPrep® high abundance terpene mix is a high concentration analytical standard for terpene analysis that contains the most abundant terpenes found in cannabis products. Terpenes are the common term for a large group of compounds that contribute to the flavor and smell of botanicals. The diversity of terpenes and terpenoids is recognized by the range of scents and flavors they produce. The terpene abundances can differ greatly between the strains of cannabis, from major terpenes, such as myrcene, often found in percent levels, to the minor terpenes, such as geraniol, mostly found in ppm levels or smaller. Terpene analysis plays a critical role in the development and QC of cannabis products as it provides a quantitative measure of the product’s flavor and scent profile. Terpene testing and profiling can be difficult since the range of abundance across the profile can vary drastically. A CRM (Certified Reference Material) can provide a basis for more accurate testing results by taking into consideration the natural differences in concentration amongst the various compounds in a terpene profile.
Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all the common contaminants such as pesticide residues, residual solvents, and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.
Hazardous; Methanol; Class 3, 6.1, Packing Group II
Flammable, Acute Toxicity, Health Hazard, Skin Sensitivity; UN1230
Certifications: Supplier Certificate of Analysis (CoA) is included with product shipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty