Would you like to log in again?
Claritas PPT® Grade ICP-MS Instrument Check Standard 3, SPEX CertiPrep
Supplier: SPEX CertiPrep
Claritas PPT®
CL-ICS-3
SXCL-ICS-3EA 246.1 USD InStock
SXCL-ICS-3
Claritas PPT® Grade ICP-MS Instrument Check Standard 3, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Claritas PPT® multi-element standards for ICP-MS
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.
Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty