Would you like to log in again?
ICP-MS Spike Sample 3, SPEX CertiPrep
Supplier: SPEX CertiPrep
CL-SPIKE-3
SXCL-SPIKE-3EA 485.67 USD InStock
SXCL-SPIKE-3
ICP-MS Spike Sample 3, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
CLP spike standard for ICP & ICP-MS.
Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.
The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by SPEX CertiPrep for convenience of use and stability.
US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentrationlevels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
- Exhibit C, Inorganic Target Analyte List (TAL)
- Exhibit D, Analytical Methods
- Exhibit E, QA/QC Requirements
CLP ILM 05.2 Standards for ICP-MS
Spike standard, CL-SPIKE-3, provides all of the analytes required for the ICP and AA spike.
Aluminum200 µg/mL
Barium200 µg/mL
Cobalt50 µg/mL
Manganese50 µg/mL
Nickel50 µg/mL
Vanadium50 µg/mL
Zinc50 µg/mL
Copper25 µg/mL
Chromium20 µg/mL
Antimony10 µg/mL
Beryllium5 µg/mL
Cadmium5 µg/mL
Silver5 µg/mL
Thallium5 µg/mL
Arsenic4 µg/mL
Lead2 µg/mL
Selenium1 µg/mL
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty