Would you like to log in again?
Claritas PPT® Grade Beryllium Standards for ICP-MS, SPEX CertiPrep
Supplier: SPEX CertiPrep
Claritas PPT®
CLBE2-2M CLBE2-2Y CLBE2-1BY
SXCLBE2-2YEA 179.79 USD InStock
SXCLBE2-2Y SXCLBE2-2M SXCLBE2-1BY
Claritas PPT® Grade Beryllium Standards for ICP-MS, SPEX CertiPrep
Standards Cationic Standards Beryllium Standards
Single element standards for ICP-MS.
  • Made with an acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)
Claritas PPT® Grade CRMs are designed for ICP and ICP-MS analysis. They are available in single and multi-element solutions. The standards are set at 1 µg/ml, 100 µg/ml, or 1,000 µg/ml and packaged in 30 ml and 125 ml bottles to minimize contamination. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.
Certifications: Certified to DQS to ISO 9001:2015
Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty