Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 2%, Claritas PPT® Grade for calibration blank in ICP

Supplier: SPEX CERTIPREP LLC
Synonyms: Muriatic acidHydrochloric acid solution

CLBLK-HCL
SXCLBLK-HCLEA 106.02 USD InStock
SXCLBLK-HCL
Hydrochloric acid 2%, Claritas PPT® Grade for calibration blank in ICP
Hydrochloric acid
Matrix blank.

May be used to dilute your multi-element standards or can be run directly as a blank to establish your base line. Do not use any acid or water as a diluent if you are not certain of its purity.


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty