Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Claritas PPT® Grade Nickel Standards for ICP-MS, SPEX CertiPrep

Supplier: SPEX CertiPrep

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Claritas PPT®
CLNI2-2Y CLNI2-1BY CLNI2-2M
SXCLNI2-1BYEA 114.67 USD InStock
SXCLNI2-1BY SXCLNI2-2M SXCLNI2-2Y
Claritas PPT® Grade Nickel Standards for ICP-MS, SPEX CertiPrep
Standards Cationic Standards Nickel Standards
Single element standards for ICP-MS.

  • Made with an acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)

Claritas PPT® Grade CRMs are designed for ICP and ICP-MS analysis. They are available in single and multi-element solutions. The standards are set at 1 µg/ml, 100 µg/ml, or 1,000 µg/ml and packaged in 30 ml and 125 ml bottles to minimize contamination. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.

Certifications: Certified to DQS to ISO 9001:2015
Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty