Would you like to log in again?
Internal Standard, 4,000 µg/mL in Methylene Chloride for US EPA Methods 625, 8270 and CLP Series Methods, SPEX CertiPrep
Supplier: SPEX CertiPrep
CLPS-I
SXCLPS-IEA 118 USD InStock
SXCLPS-I
Internal Standard, 4,000 µg/mL in Methylene Chloride for US EPA Methods 625, 8270 and CLP Series Methods, SPEX CertiPrep
Standards Environmental Analysis Standards
Internal Standard for US EPA Methods 625, 8270 and CLP Series Methods, 4,000 µg/mL (4,000 ppm) in Methylene Chloride, 1 mL
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 625, 8270 and CLP Series Methods
  • 4,000 ppm Internal Standard in methylene chloride matrix
Analytes: Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, Phenanthrene-d10
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty