Would you like to log in again?
Calcium (Ca2+) Single-Element Ion Cation Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
CS-CA2-2Y
SXCS-CA2-2YEA 87.2 USD InStock
SXCS-CA2-2Y
Calcium (Ca2+) Single-Element Ion Cation Standard, SPEX CertiPrep
Standards Cationic Standards Calcium Standards
Single-element standards for ion chromatography and ion selective electodes.
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty