Would you like to log in again?
Assurance® Grade Mixed Calibration Standard 2 for AA and ICP, SPEX CertiPrep
Supplier: SPEX CertiPrep
Assurance®
MIXSTD2-100 MIXSTD2-500
SXMIXSTD2-100EA 236.49 USD InStock
SXMIXSTD2-100 SXMIXSTD2-500
Assurance® Grade Mixed Calibration Standard 2 for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Assurance® mixed multi-element standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
AA & ICP Calibration Standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.
For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty