Would you like to log in again?
Assurance® Grade Trace Metals III for AA & ICP, SPEX CertiPrep
Supplier: SPEX CertiPrep
Assurance®
MN-6-500 MN-6
SXMN-6EA 244.36 USD InStock
SXMN-6 SXMN-6-500
Assurance® Grade Trace Metals III for AA & ICP, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Groundwater and wastewater pollution control check standards. Assurance® mixed multi-element standards for AA & ICP.
Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.
May be used either as standards or as a means to check the individual analysts accuracy and precision.
Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty