Would you like to log in again?
Certified pH Buffer Standards, SPEX CertiPrep
Supplier: SPEX CertiPrep
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH-BUFF9-500 PH-BUFF3-500 PH-BUFF8-500 PH-BUFF2-500 PH-BUFF4-500 PH-BUFF11-500 PH-BUFF7-500 PH-BUFF5-500 PH-BUFF6-500 PH-BUFF12-500 PH-BUFF10-500
SXPH-BUFF2-500EA 69.97 USD InStock
SXPH-BUFF2-500 SXPH-BUFF7-500 SXPHBUFF12-500 SXPHBUFF11-500 SXPH-BUFF6-500 SXPH-BUFF8-500 SXPH-BUFF9-500 SXPHBUFF10-500 SXPH-BUFF4-500 SXPH-BUFF5-500 SXPH-BUFF3-500
Certified pH Buffer Standards, SPEX CertiPrep
Buffers pH Calibration Buffers
Certified pH buffers are made using high purity starting materials and ASTM Type I Water, and are thoroughly analyzed for certified value and homogeneity. Buffers are accurate, premium grade for calibration and quality control checks.
  • Conveniently packaged in 500 ml LDPE bottles.
Certifications: SPEX CertiPrep pH Buffers are accurate and certified to ISO/IEC 17025:2005 and ISO 17034:2016 requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty