Would you like to log in again?
Pipette Washer and Dryer, SPEX CertiPrep®
Supplier: SPEX CertiPrep
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIPWASH-1 PIPPUMP-230V PIPPUMP-115V PIPWASH-1 PIPBASIN-1 PIPBASIN-1
SXPIPWASH-1EA 1277.71 USD InStock
CA11023-394 SXPIPWASH-1 SXPIPPUMP-230V CA11023-392 SXPIPBASIN-1 SXPIPPUMP-115V
Pipette Washer and Dryer, SPEX CertiPrep®
Pipettes Pipette Washers
The Pipette Washer and Dryer was designed by SPEX CertiPrep® chemists to take the hassle out of washing Class A volumetric pipettes.
  • Lightweight and compact, the washer/dryer fits within a sink or atop lab bench
  • Durable polyethylene construction
  • Convenient, independent on/off valves control flow
  • Transparent door closes to prevent splashing when using acids in water
  • 23 cone-shaped, plastic pipette holders accommodate pipette sizes ranging from 0.5 to 250 ml
  • Optional pump and basin available separately
This easy to use device requires users to simply insert up to 23 pipettes at a time, close the door, and attach tubing to the wash or rinse line. The washer can also be used with the washer basin and pump to circulate wash or rinse solution through the pipettes. The solution shoots out of the pipette tip, reflects off the ceiling portion of the washer, and rains a shower down over the outside of the pipettes. This process effectively cleans both the interior and exterior of the pipettes. To dry the interior of the pipettes the line is connected to a vacuum source and air is pulled in through the pipette tips until all interiors are dry.
W×D×H= 1×1×3"
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty