Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Assurance® Grade Nickel Standards for AA and ICP, SPEX CertiPrep

Supplier: SPEX CertiPrep

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Assurance®
PLNI2-2T PLNI2-2M PLNI2-3Y PLNI2-2X PLNI2-2Y PLNI2-3X
SXPLNI2-2MEA 37.69 USD InStock
SXPLNI2-2M SXPLNI2-2T SXPLNI2-2Y SXPLNI2-3Y SXPLNI2-2X SXPLNI2-3X
Assurance® Grade Nickel Standards for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Nickel Standards
Single-element standards for AA & ICP.

  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)

AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty