Would you like to log in again?
Assurance® Grade Tellurium Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CertiPrep
Assurance®
PLTE4-2X PLTE4-2M PLTE4-2Y
SXPLTE4-2XEA 183.84 USD InStock
SXPLTE4-2X SXPLTE4-2Y SXPLTE4-2M
Assurance® Grade Tellurium Standards for AA and ICP, SPEX CertiPrep
Standards Cationic Standards Tellurium Standards
Single-element standards for AA & ICP.
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)
AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.
Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty