Would you like to log in again?
Diisodecyl phthalate Single-Component Organic Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
S-4464 CRM-PE-DIDP
SXS-4464EA 80.35 USD InStock
SXS-4464 SXCRM-PE-DIDP
Diisodecyl phthalate Single-Component Organic Standard, SPEX CertiPrep
Standards Organic Standards
Consumer safety standards, phthalates in polyethylene QC standards.
Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty