Would you like to log in again?
Multi-Element Organometallic Oil Standards, 21 Elements, SPEX CertiPrep
Supplier: SPEX CertiPrep
S21-900Y S21-100Z S21-900Z S21-100Y S21-300Y S21-500Y S21-300Z S21-500Z
SXS21-900ZEA 421.87 USD InStock
SXS21-900Z SXS21-900Y SXS21-100Z SXS21-100Y SXS21-300Y SXS21-300Z SXS21-500Z SXS21-500Y
Multi-Element Organometallic Oil Standards, 21 Elements, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
SPEX CertiPrep presents a comprehensive offering of Organometallic Oil Standards. The applications of these standards include:
  • Crude oils
  • Additive metals
  • Environmental monitoring
  • Petrochemical testing
  • Pharmaceuticals
  • Food processing
  • Sulfur in diesel fuel
The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.
- Choice of over 35 single-elements at 1,000 or 5,000 μg/g
- Popular multi-element blends of 23, 21, 12, or 5 elements
- Clear, transparent matrix
- Guaranteed stable and accurate
Certificate of Analysis with every solution
Certifications: Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty