Would you like to log in again?
Assurance® Grade Arsenic +5, Speciation Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
Assurance®
SPEC-AS5M SPEC-AS5
SXSPEC-AS5MEA 37.69 USD InStock
SXSPEC-AS5M SXSPEC-AS5
Assurance® Grade Arsenic +5, Speciation Standard, SPEX CertiPrep
Standards Cationic Standards Arsenic Standards
Speciation analysis has become common in many fields, including environmental, food and pharmaceutical testing labs. To analyze species within a sample requires Certified Reference Materials (CRMs) for sample verification and method validation.
Many speciation standards are available in today’s market, however, most of them are not certified or analyzed with a state-of-the-art ICP, ICP-MS or LC-ICP-MS. SPEX CertiPrep offers a wide variety of speciation standards, certified to the strictest ISO 17034 guidelines, and tested on LC-ICP-MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty