Would you like to log in again?
USP <561> Standards, 100 µg/ml, Spex CertiPrep®
Supplier: SPEX CertiPrep
USP-561-F USP-561-D USP-561-E USP-561-A USP-561-C USP-561-G USP-561-B
SXUSP-561-GEA 581.59 USD InStock
SXUSP-561-G SXUSP-561-F SXUSP-561-E SXUSP-561-D SXUSP-561-C SXUSP-561-B SXUSP-561-A
USP <561> Standards, 100 µg/ml, Spex CertiPrep®
Standards Organic Standards
Tests and specifications for the safety of botanical products.
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
The United States Pharmacopeia (USP) creates standards for the monitoring of pharmaceutical and botanical products. General Chapter <561> Articles of Botanical Origin, outlines tests and specifications for the safety of botanical products. The chapter outlines sampling procedures and tests for general characteristics, such as for moisture, ash, starch and fiber. The chapter also describes methods for extractables and volatile compounds. In the interests of safety, USP <561> has procedures for the testing of mycotoxins and designates targeted pesticides. SPEX CertiPrep provides standards for testing of mycotoxins and pesticides under the General Chapter <561>.
Certifications: Supplier Certificate of Analysis (CoA) is included with product shipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty