Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
5 Organic Volatile Impurities,Class 1 Solvents, USP 467 Pharmaceutical Residual Solvent Standards, SPEX CertiPrep
Supplier: SPEX CertiPrep
USP-RS-C1
SXUSP-RS-C1EA 52.39 USD InStock
SXUSP-RS-C1
5 Organic Volatile Impurities,Class 1 Solvents, USP 467 Pharmaceutical Residual Solvent Standards, SPEX CertiPrep
Standards Environmental Analysis Standards
Consumer safety standards. Pharmaceutical residual solvent standards.
In the pharmaceutical industry, the guidelines set by the International Conference on Harmonization (ICH), and by United States Pharmacopeia (USP) and European Pharmacopeia (EP), mandate that manufacturing solvents have to be regulated due to their toxic and/or environmentally hazardous nature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty