Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.5%

Supplier: TCI America

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

A0060-500ML A0060-25ML
TCA0060-025MLEA 17.08 USD InStock
TCA0060-025ML TCA0060-500ML
Acetonitrile ≥99.5%
Acetonitrile
CAS Number: 75-05-8
MDL Number: MFCD00001878
Molecular Formula: C2H3N
Molecular Weight: 41.05
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Melting point (°C): -46
Flash Point (°C): 10
Specific Gravity (20/20): 0.78

Other Product Names / Synonyms: Methyl Cyanide, ACN


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C
Melting Pt: –46 °C
Density: 0.78 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/75/02/28927502.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.5% (GC)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty