Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-4-hydroxybenzoic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A0859-5G A0859-25G
TCA0859-005GEA 114.62 USD InStock
TCA0859-005G TCA0859-25G
3-Amino-4-hydroxybenzoic acid ≥97.0% (by HPLC, titration analysis)
3-Amino-4-hydroxybenzoic acid
CAS Number: 1571-72-8
MDL Number: MFCD00007697
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 197
Lambda max.: 308 nm (H2O (pH 11))


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 197 °C
Storage Temperature: Ambient
MDL Number: MFCD00007697
CAS Number: 1571-72-8
EINECS: 216-390-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/04/28928604.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty