Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥97.0% free radical

Supplier: TCI America
Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl

A1343-5G A1343-1G
TCA1343-005GEA 308.62 USD InStock
TCA1343-005G TCA1343-001G
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥97.0% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)
CAS Number: 14691-88-4
MDL Number: MFCD00006479
Molecular Formula: C9H19N2O
Molecular Weight: 171.26
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 98
Freezing point (°C): 39
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Amino-TEMPO


Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Boiling Pt: 98 °C
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/56/28929256.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty