Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) ≥98.0% free radical

Supplier: TCI America
Synonyms: 4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl

A1348-25G A1348-5G
TCA1348-005GEA 51.23 USD InStock
TCA1348-005G TCA1348-25G
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) ≥98.0% free radical
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-Acetamido-TEMPO, free radical)
[Catalyst for Oxidation]
CAS Number: 14691-89-5
MDL Number: MFCD00043593
Molecular Formula: C11H21N2O2
Molecular Weight: 213.30
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 143
Flash Point (°C): 198

Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Acetamido-TEMPO


Formula: C₁₁H₂₁N₂O₂
MW: 213.3 g/mol
Melting Pt: 143 °C
Flash Pt: 198 °C
MDL Number: MFCD00043593
CAS Number: 14691-89-5
EINECS: 423-840-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/65/28929265.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty