Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SGPG(HNK-1) Mouse Monoclonal Antibody [clone: NGR50]

Supplier: TCI
A2706-VIAL
TCA2706-VIALEA 771 USD InStock
TCA2706-VIAL
Anti-SGPG(HNK-1) Mouse Monoclonal Antibody [clone: NGR50]
Antibodies
Purified mouse monoclonal antibody/IgG2a, κ -chain

Type: Primary
Antigen: SGPG(HNK-1)
Clonality: Monoclonal
Clone: NGR50
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty