Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium benzenesulfonate ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Benzenesulphonic acid, sodium saltBenzenesulfonic acid, sodium saltBenzenesulfonic acid Sodium SaltBenzenesulphonic acid Sodium Salt

B0035-25G B0035-500G B0035-100G
TCB0035-500GEA 216.13 USD InStock
TCB0035-500G TCB0035-025G TCB0035-100G
Sodium benzenesulfonate ≥96.0% (by titrimetric analysis)
Sodium benzenesulfonate
CAS Number: 515-42-4
MDL Number: MFCD00065179
Molecular Formula: C6H6O3S
Molecular Weight: 180.15
Purity/Analysis Method: 96.0% (T)
Form: Crystal

Other Product Names / Synonyms: Benzenesulfonic Acid Sodium Salt


Formula: C₆H₆O₃S
MW: 180.15 g/mol
MDL Number: MFCD00065179
CAS Number: 515-42-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/09/28932209.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty