Synonyms:
2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride
B0457-500G
B0457-25G
TCB0457-025GEA
28.4
USD
InStock
TCB0457-025G
TCB0457-500G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)
Girard's reagent T (Carbazoylmethyltrimethylammonium chloride)
CAS Number: 123-46-6
MDL Number: MFCD00012009
Molecular Formula: C5H14ClN3O
Molecular Weight: 167.64
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 190
Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: Betaine Hydrazide Hydrochloride, (Hydrazinocarbonylmethyl)trimethylammonium Chloride, Trimethylacetohydrazideammonium Chloride