Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

B0457-500G B0457-25G
TCB0457-025GEA 28.4 USD InStock
TCB0457-025G TCB0457-500G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)
Girard's reagent T (Carbazoylmethyltrimethylammonium chloride)
CAS Number: 123-46-6
MDL Number: MFCD00012009
Molecular Formula: C5H14ClN3O
Molecular Weight: 167.64
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 190

Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: Betaine Hydrazide Hydrochloride, (Hydrazinocarbonylmethyl)trimethylammonium Chloride, Trimethylacetohydrazideammonium Chloride


Formula: C₅H₁₄ClN₃O
MW: 167.64 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00012009
CAS Number: 123-46-6
EINECS: 204-629-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/24/22/28932422.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty