Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Bis(5-phenyloxazol-2-yl)benzene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning
Synonyms: POPOP

B0509-1G B0509-25G B0509-5G
TCB0509-5GEA 83.2 USD InStock
TCB0509-5G TCB0509-001G TCB0509-025G
1,4-Bis(5-phenyloxazol-2-yl)benzene ≥98.0% (by HPLC, total nitrogen)
1,4-Bis(5-phenyloxazol-2-yl)benzene
[for scintillation spectrometry]
CAS Number: 1806-34-4
MDL Number: MFCD00005309
Molecular Formula: C24H16N2O2
Molecular Weight: 364.40
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 245

Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: 1,4-Di(5-phenyl-2-oxazolyl)benzene, POPOP


Formula: C₂₄H₁₆N₂O₂
MW: 364.4 g/mol
Melting Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00005309
CAS Number: 1806-34-4
EINECS: 217-304-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/24/61/28932461.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty