Synonyms:
POPOP
B0509-1G
B0509-25G
B0509-5G
TCB0509-5GEA
83.2
USD
InStock
TCB0509-5G
TCB0509-001G
TCB0509-025G
1,4-Bis(5-phenyloxazol-2-yl)benzene ≥98.0% (by HPLC, total nitrogen)
1,4-Bis(5-phenyloxazol-2-yl)benzene
[for scintillation spectrometry]
CAS Number: 1806-34-4
MDL Number: MFCD00005309
Molecular Formula: C24H16N2O2
Molecular Weight: 364.40
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 245
Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: 1,4-Di(5-phenyl-2-oxazolyl)benzene, POPOP