Synonyms:
Diethyl ether boron trifluoride Boron trifluoride diethyl ether Boron trifluoride diethyl etherate Boron trifluoride ethyl etherate
B0527-500ML
B0527-100ML
B0527-25ML
TCB0527-100MLEA
50.29
USD
InStock
TCB0527-100ML
TCB0527-25ML
TCB0527-500ML
Boron trifluoride diethyl ether complex ≥98% (by gravimetric analysis)
Boron trifluoride diethyl ether complex
CAS Number: 109-63-7
Molecular Formula: C4H10BF3O
Molecular Weight: 141.93
Form: Liquid
Color: Colorless
Product Categories: Analytical Science, NMR Spectrometry, Bioscience, Sugars, Synthetic Organic Chemistry, Carbonyl Olefination, Functional Group Transformation and Introducing Reaction, Multicomponent Reaction (MCR)