Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boron trifluoride diethyl ether complex ≥98% (by gravimetric analysis)

Supplier: TCI America
Danger
Synonyms: Diethyl ether boron trifluoride Boron trifluoride diethyl ether Boron trifluoride diethyl etherate Boron trifluoride ethyl etherate

B0527-500ML B0527-100ML B0527-25ML
TCB0527-100MLEA 50.29 USD InStock
TCB0527-100ML TCB0527-25ML TCB0527-500ML
Boron trifluoride diethyl ether complex ≥98% (by gravimetric analysis)
Boron trifluoride diethyl ether complex
CAS Number: 109-63-7
Molecular Formula: C4H10BF3O
Molecular Weight: 141.93
Form: Liquid
Color: Colorless

Product Categories: Analytical Science, NMR Spectrometry, Bioscience, Sugars, Synthetic Organic Chemistry, Carbonyl Olefination, Functional Group Transformation and Introducing Reaction, Multicomponent Reaction (MCR)


Formula: C₄H₁₀BF₃O
MW: 141.93 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00013194
CAS Number: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8,I
Merck Index: 13,01340
https://us-legacy.vwr.com/stibo/web/std.lang.all/24/75/28932475.jpg
Order Now
Specification Test Results
Purity(Gravimetric Method) 98.0 to 102.0 %
Specific gravity (20/20) 1.1250 to 1.1320
Refractive index n20/D 1.3430 to 1.3470

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty