Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Bromo-2-naphthol ≥98.0%

Supplier: TCI America
Synonyms: 6-Bromo-2-hydroxynaphthalene 2-Bromo-6-hydroxynaphthalene

B0621-25G B0621-100G
TCB0621-100GEA 115.47 USD InStock
TCB0621-100G TCB0621-025G
6-Bromo-2-naphthol ≥98.0%
6-Bromo-2-naphthol
CAS Number: 15231-91-1
MDL Number: MFCD00004081
Molecular Formula: C10H7BrO
Molecular Weight: 223.07
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 129


Formula: C₁₀H₇BrO
Boiling Pt: 200…205 °C (20 mmHg)
Melting Pt: 129 °C
MDL Number: MFCD00004081
CAS Number: 15231-91-1
EINECS: 239-279-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/52/28932552.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty