Synonyms:
Benzene-[²H₆]
B0840-25ML
TCB0840-25MLEA
253.15
USD
InStock
TCB0840-25ML
Benzene-[D₆] (99.6% D)
Benzene-[D6]
CAS Number: 1076-43-3
MDL Number: MFCD00003010
Molecular Formula: C6D6
Molecular Weight: 84.15
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Melting point (°C): 7
Flash Point (°C): -11
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Hexadeuterobenzene