Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzofuroxan ≥98.0%

Supplier: TCI America
Synonyms: 2,1,3-Benzoxadiazole 1-OxideBenzofurazan 1-Oxide

B1316-25G B1316-500G
TCB1316-025GEA 51.77 USD InStock
TCB1316-025G TCB1316-500G
Benzofuroxan ≥98.0%
Benzofuroxan
CAS Number: 480-96-6
MDL Number: MFCD00005807
Molecular Formula: C6H4N2O2
Molecular Weight: 136.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 69

Other Product Names / Synonyms: Benzofurazan 1-Oxide, 2,1,3-Benzoxadiazole 1-Oxide


Formula: C₆H₄N₂O₂
MW: 136.11 g/mol
Melting Pt: 69 °C
MDL Number: MFCD00005807
CAS Number: 480-96-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/40/28933140.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty