Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-tert-Butyl-ɑ-phenylnitrone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: N-Benzylidene-tert-butylamine N-OxidePBNN-tert-Butyl-α-phenylnitrone

B1701-5G B1701-1G
TCB1701-005GEA 221.21 USD InStock
TCB1701-005G TCB1701-001G
N-tert-Butyl-ɑ-phenylnitrone ≥98.0% (by HPLC, titration analysis)
N-tert-Butyl-alpha-phenylnitrone
CAS Number: 3376-24-7
MDL Number: MFCD00008799
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 74

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: N-Benzylidene-tert-butylamine N-Oxide, PBN


Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Melting Pt: 74 °C
MDL Number: MFCD00008799
CAS Number: 3376-24-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/55/28933455.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty