Synonyms:
N-Benzylidene-tert-butylamine N-OxidePBNN-tert-Butyl-α-phenylnitrone
B1701-5G
B1701-1G
TCB1701-005GEA
221.21
USD
InStock
TCB1701-005G
TCB1701-001G
N-tert-Butyl-ɑ-phenylnitrone ≥98.0% (by HPLC, titration analysis)
N-tert-Butyl-alpha-phenylnitrone
CAS Number: 3376-24-7
MDL Number: MFCD00008799
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 74
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: N-Benzylidene-tert-butylamine N-Oxide, PBN