Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: N-tert-Butyl-alpha-(4-pyridyl)nitrone N'-OxidePOBNN-(4-Pyridylmethylene)-tert-butylamine N,N'-Dioxidealpha-(4-Pyridyl N-Oxide)-N-tert-butylnitrone

B1735-1G B1735-5G
TCB1735-001GEA 227.26 USD InStock
TCB1735-001G TCB1735-5G
N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone ≥98.0% (by HPLC, titration analysis)
N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone
CAS Number: 66893-81-0
MDL Number: MFCD00041022
Molecular Formula: C10H14N2O2
Molecular Weight: 194.23
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185
Storage Temperature: 0-10°C

Product Categories: Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: N-tert-Butyl-alpha-(4-pyridyl)nitrone N'-Oxide, POBN, N-(4-Pyridylmethylene)-tert-butylamine N,N'-Dioxide, alpha-(4-Pyridyl N-Oxide)-N-tert-butylnitrone


Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00041022
CAS Number: 66893-81-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/83/28933483.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty