Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzyloxy-4-bromobenzene ≥98.0%

Supplier: TCI America
Synonyms: 1-Bromo-4-(phenylmethoxy)benzene4-Benzyloxybromobenzene

B2127-25G B2127-5G
TCB2127-025GEA 105.56 USD InStock
TCB2127-025G TCB2127-5G
1-Benzyloxy-4-bromobenzene ≥98.0%
1-Benzyloxy-4-bromobenzene
CAS Number: 6793-92-6
MDL Number: MFCD00028016
Molecular Formula: C13H11BrO
Molecular Weight: 263.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 62

Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Benzyl 4-Bromophenyl Ether, 4-Benzyloxybromobenzene


Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00028016
CAS Number: 6793-92-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/38/99/28933899.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty