Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: N-Boc-4-fluoro-D-phenylalanineBoc-D-Phe(4-F)-OH

B3303-1G B3303-5G
TCB3303-5GEA 168.49 USD InStock
TCB3303-5G TCB3303-1G
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
CAS Number: 57292-45-2
MDL Number: MFCD00079669
Molecular Formula: C14H18FNO4
Molecular Weight: 283.30
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 87
Specific rotation [a]20/D: -20 deg (C=1, EtOH)

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-4-fluoro-D-phenylalanine, Boc-D-Phe(4-F)-OH


Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00079669
CAS Number: 57292-45-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/46/98/28934698.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty